4-Bromobenzo[b]thiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

047307-1G
101879-434EA 698.46 USD
101879-434
4-Bromobenzo[b]thiophene-2-carboxylic acid ≥95%
4-Bromobenzo[b]thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅BrO₂S
MW: 257.1038 g/mol
Melting Pt: 265 °C
MDL Number: MFCD03407342
CAS Number: 5194-37-6

Order Now

Specification Test Results

Melting point 265°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR