4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047301-500MG 047301-5G
101879-400EA 375.38 USD
101879-400 101879-404
4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde ≥95%
4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde

Hazard: Irritant

Formula: C₂₃H₂₂O₄
MW: 362.43 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD08689784
CAS Number: 937602-07-8

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR