3-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047300-1G
101879-396EA 467.69 USD
101879-396
3-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde ≥95%
3-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde

Hazard: Irritant

Formula: C₂₃H₂₂O₄
MW: 362.43 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD08689783
CAS Number: 937602-05-6

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR