1-(Benzyloxy)-4-(3-bromopropoxy)benzene ≥95%

Supplier: Matrix Scientific

047294-1G
101879-360EA 350.77 USD
101879-360
1-(Benzyloxy)-4-(3-bromopropoxy)benzene ≥95%
1-(Benzyloxy)-4-(3-bromopropoxy)benzene

Hazard: Irritant

Formula: C₁₆H₁₇BrO₂
Melting Pt: 61…63 °C
MDL Number: MFCD08689778
CAS Number: 80199-92-4

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR