8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride ≥95%

Supplier: Matrix Scientific

047291-5G 047291-500MG
101879-344EA 806.15 USD
101879-344 101879-340
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride ≥95%
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂
MW: 222.72 g/mol
Melting Pt: 270…271 °C
MDL Number: MFCD07781100
CAS Number: 57933-28-5

Order Now

Specification Test Results

Melting point 270-271°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR