Methyl-3-[2-(4-formylphenoxy)ethoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
047288-1G
101879-324EA
467.69
USD
101879-324
Methyl-3-[2-(4-formylphenoxy)ethoxy]benzenecarboxylate ≥95%
Methyl-3-[2-(4-formylphenoxy)ethoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₇H₁₆O₅ MW: 300.31 g/mol Melting Pt: 85…87 °C |
MDL Number:
MFCD08689774 CAS Number: 937601-95-1 |
Specification Test Results
Melting point | 85-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...