3-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047275-1G
101879-248EA 350.77 USD
101879-248
3-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde ≥95%
3-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde

Hazard: Irritant

Formula: C₂₂H₂₀O₄
MW: 348.4 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD08689764
CAS Number: 937601-86-0

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR