Methyl-4-[2-(4-formylphenoxy)ethoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
047271-1G
101879-224EA
350.77
USD
101879-224
Methyl-4-[2-(4-formylphenoxy)ethoxy]benzenecarboxylate ≥95%
Methyl-4-[2-(4-formylphenoxy)ethoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₇H₁₆O₅ MW: 300.31 g/mol Melting Pt: 107…109 °C |
MDL Number:
MFCD08689761 CAS Number: 866003-00-1 |
Specification Test Results
Melting point | 107-109°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...