1-(Benzyloxy)-4-(2-bromoethoxy)benzene ≥95%
Supplier: Matrix Scientific
047263-5G
047263-500MG
101879-176EA
181.54
USD
101879-176
101879-180
1-(Benzyloxy)-4-(2-bromoethoxy)benzene ≥95%
1-(Benzyloxy)-4-(2-bromoethoxy)benzene
Hazard: Irritant
Formula:
C₁₅H₁₅BrO₂ MW: 307.19 g/mol Melting Pt: 85…87 °C |
MDL Number:
MFCD08689757 CAS Number: 3351-59-5 |
Specification Test Results
Melting point | 85-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...