Methyl-6-amino-5-nitro-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

047262-500MG
101879-172EA 1109.23 USD
101879-172
Methyl-6-amino-5-nitro-2-pyridinecarboxylate ≥95%
Methyl-6-amino-5-nitro-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₇H₇N₃O₄
MW: 197.15 g/mol
Melting Pt: 191…193 °C
MDL Number: MFCD12827782
CAS Number: 538372-32-6

Order Now

Specification Test Results

Melting point 191-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR