6-(Dimethylamino)nicotinonitrile ≥95%

Supplier: Matrix Scientific

047260-1G
101879-168EA 350.77 USD
101879-168
6-(Dimethylamino)nicotinonitrile ≥95%
6-(Dimethylamino)nicotinonitrile

Hazard: Irritant

Formula: C₈H₉N₃
Melting Pt: 66…68 °C
MDL Number: MFCD08272099
CAS Number: 154924-17-1

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR