2-(Methylamino)nicotinonitrile ≥95%

Supplier: Matrix Scientific

047254-1G
101879-132EA 232.31 USD
101879-132
2-(Methylamino)nicotinonitrile ≥95%
2-(Methylamino)nicotinonitrile

Hazard: Irritant

Formula: C₇H₇N₃
MW: 133.1506 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD08689752
CAS Number: 52583-87-6

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR