1-(4-Formylphenyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047252-1G
101879-120EA 350.77 USD
101879-120
1-(4-Formylphenyl)-4-piperidinecarboxylic acid ≥95%
1-(4-Formylphenyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Melting Pt: 258…261 °C
MDL Number: MFCD08689751
CAS Number: 727396-60-3

Order Now

Specification Test Results

Melting point 258-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR