Methyl-3-amino-4-fluorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-fluorobenzoate

047241-1G
101879-066EA 32.31 USD
101879-066
Methyl-3-amino-4-fluorobenzoate ≥95%
Methyl 3-amino-4-fluorobenzoate

Hazard: Irritant

Formula: C₈H₈FNO₂
MW: 169.1500 g/mol
Melting Pt: 66…69 °C
MDL Number: MFCD08689743
CAS Number: 369-26-6

Order Now

Specification Test Results

Melting point 66-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR