1-(6-Bromo-2-pyridinyl)-3-(dimethylamino)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

047216-1G
101878-964EA 350.77 USD
101878-964
1-(6-Bromo-2-pyridinyl)-3-(dimethylamino)-2-propen-1-one ≥95%
1-(6-Bromo-2-pyridinyl)-3-(dimethylamino)-2-propen-1-one

Hazard: Irritant

Formula: C₁₀H₁₁BrN₂O
MW: 255.12 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD08689738
CAS Number: 638197-51-0

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR