(R)-(+)-STERICOL ≥99%
Supplier: Matrix Scientific
047214-1G
101878-952EA
350.77
USD
101878-952
(R)-(+)-STERICOL ≥99%
(R)-(+)-STERICOL
Hazard: Irritant
Formula:
C₁₇H₂₈O Melting Pt: 88 °C |
MDL Number:
MFCD09836214 CAS Number: 181531-14-6 |
Specification Test Results
Melting point | 88°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...