Methyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carboxylate ≥95%

Supplier: Matrix Scientific

047210-500MG
101878-934EA 1109.23 USD
101878-934
Methyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carboxylate ≥95%
Methyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂O₂
Melting Pt: 97…98 °C
MDL Number: MFCD12827781
CAS Number: 1186405-14-0

Order Now

Specification Test Results

Melting point 97-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR