2,6-Dichloro-3-pyridinemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloro-3-(hydroxymethyl)pyridine(2,6-Dichloropyridin-3-yl)methanol

047209-1G
101878-930EA 60 USD
101878-930
2,6-Dichloro-3-pyridinemethanol ≥95%
2,6-Dichloro-3-pyridinemethanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.012 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD11036367
CAS Number: 55304-90-0

Order Now

Specification Test Results

Melting point 67-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR