5-Bromo-6-methyl-2-(methylamino)-4-(trifluoromethyl)nicotinonitrile ≥95%
Supplier: Matrix Scientific
047208-1G
047208-500MG
101878-928EA
698.46
USD
101878-928
101878-926
5-Bromo-6-methyl-2-(methylamino)-4-(trifluoromethyl)nicotinonitrile ≥95%
5-Bromo-6-methyl-2-(methylamino)-4-(trifluoromethyl)nicotinonitrile
Hazard: Irritant
Formula:
C₉H₇BrF₃N₃ Melting Pt: 171…173 °C |
MDL Number:
MFCD12498720 CAS Number: 1171919-20-2 |
Specification Test Results
Melting point | 171-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...