Methyl-2-(5-bromo-2-pyrimidinyl)-2-cyanoacetate ≥95%
Supplier: Matrix Scientific
047203-1G
047203-500MG
101878-912EA
467.69
USD
101878-912
101878-910
Methyl-2-(5-bromo-2-pyrimidinyl)-2-cyanoacetate ≥95%
Methyl-2-(5-bromo-2-pyrimidinyl)-2-cyanoacetate
Hazard: Irritant
Formula:
C₈H₆BrN₃O₂ Melting Pt: 222…224 °C |
MDL Number:
MFCD12498717 CAS Number: 1171919-13-3 |
Specification Test Results
Melting point | 222-224°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...