Methyl-2-amino-3-chloro-5-(trifluoromethyl)isonicotinate ≥95%
Supplier: Matrix Scientific
047201-500MG
047201-1G
101878-902EA
1400
USD
101878-902
101878-900
Methyl-2-amino-3-chloro-5-(trifluoromethyl)isonicotinate ≥95%
Methyl-2-amino-3-chloro-5-(trifluoromethyl)isonicotinate
Hazard: Irritant
Formula:
C₈H₆ClF₃N₂O₂ MW: 254.6 g/mol Melting Pt: 131…132 °C |
MDL Number:
MFCD12498715 CAS Number: 1171919-10-0 |
Specification Test Results
Melting point | 131-132°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...