N,3-Dihydroxyisonicotinamide ≥95%

Supplier: Matrix Scientific

047192-500MG 047192-1G
101878-866EA 1076.92 USD
101878-866 101878-864
N,3-Dihydroxyisonicotinamide ≥95%
N,3-Dihydroxyisonicotinamide

Hazard: Irritant

Formula: C₆H₆N₂O₃
Melting Pt: 208…210 °C
MDL Number: MFCD12827778
CAS Number: 89640-77-7

Order Now

Specification Test Results

Melting point 208-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR