6-Chloro-7-iodo-1H-pyrrolo[3,2-c]pyridine ≥95%
Supplier: Matrix Scientific
047190-500MG
101878-858EA
764.62
USD
101878-858
6-Chloro-7-iodo-1H-pyrrolo[3,2-c]pyridine ≥95%
6-Chloro-7-iodo-1H-pyrrolo[3,2-c]pyridine
Hazard: Irritant
Formula:
C₇H₄ClIN₂ Melting Pt: 245 °C |
MDL Number:
MFCD12498711 CAS Number: 1171919-03-1 |
Specification Test Results
Melting point | 245°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...