6-Chloro-2-phenyl-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

047187-500MG 047187-1G 047187-5G
101878-840EA 290.77 USD
101878-840 101878-844 101878-842
6-Chloro-2-phenyl-4-pyrimidinamine ≥95%
6-Chloro-2-phenyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₀H₈ClN₃
Melting Pt: 135…137 °C
MDL Number: MFCD12498708
CAS Number: 90799-81-8

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR