Methyl-6-chloro-3-chromanecarboxylate ≥95%
Supplier: Matrix Scientific
047180-500MG
047180-1G
047180-5G
101878-808EA
2129.23
USD
101878-808
101878-806
101878-804
Methyl-6-chloro-3-chromanecarboxylate ≥95%
Methyl-6-chloro-3-chromanecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁ClO₃ Melting Pt: 48…50 °C |
MDL Number:
MFCD04114610 CAS Number: 68281-66-3 |
Specification Test Results
Melting point | 48-50°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...