Methyl-5-(trifluoromethoxy)-1H-indazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-(trifluoromethoxy)-1H-indazole-3-carboxylate

047169-500MG
101878-754EA 1296.92 USD
101878-754
Methyl-5-(trifluoromethoxy)-1H-indazole-3-carboxylate ≥95%
Methyl-5-(trifluoromethoxy)-1H-indazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O₃
MW: 260.17 g/mol
Melting Pt: 181…182 °C
MDL Number: MFCD12498703
CAS Number: 932041-12-8

Order Now

Specification Test Results

Melting point 181-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR