4-Amino-5-iodo-2-(trifluoromethyl)benzenecarbonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Amino-5-iodo-2-(trifluoromethyl)benzonitrile
047162-500MG
047162-1G
047162-5G
101878-736EA
1384.62
USD
101878-736
101878-734
101878-732
4-Amino-5-iodo-2-(trifluoromethyl)benzenecarbonitrile ≥97%
4-Amino-5-iodo-2-(trifluoromethyl)benzenecarbonitrile
Hazard: Irritant
Formula:
C₈H₄F₃IN₂ MW: 312.03 g/mol Melting Pt: 170…172 °C |
MDL Number:
MFCD09838955 CAS Number: 852569-35-8 |
Specification Test Results
Melting point | 170-172°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...