Ethyl-4-(3-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
047142-500MG
047142-5G
047142-1G
101878-632EA
232.31
USD
101878-632
101878-630
101878-634
Ethyl-4-(3-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(3-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₄ Melting Pt: 82…85 °C |
MDL Number:
MFCD12025939 CAS Number: 219814-74-1 |
Specification Test Results
Melting point | 82-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...