1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
047132-500MG
047132-1G
101878-576EA
1163.08
USD
101878-576
101878-574
1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₀ClNO₂ Melting Pt: 73…75 °C |
MDL Number:
MFCD11553029 CAS Number: 1092352-35-6 |
Specification Test Results
Melting point | 73-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...