Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

047131-500MG 047131-1G 047131-5G
101878-572EA 229.23 USD
101878-572 101878-570 102833-148
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate ≥95%
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₄
MW: 207.19 g/mol
Melting Pt: 256…258 °C
MDL Number: MFCD07774184
CAS Number: 142166-00-5

Order Now

Specification Test Results

Melting point 256-258°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR