5-Chloro-1-(4-methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
047125-500MG
047125-1G
101878-548EA
927.69
USD
101878-548
101878-546
5-Chloro-1-(4-methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Chloro-1-(4-methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₂ClNO₂ Melting Pt: 112…114 °C |
MDL Number:
MFCD11100230 CAS Number: 1048913-54-7 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...