2-[4-(tert-Butoxycarbonyl)piperazino]-3-chloroisonicotinic acid ≥95%
Supplier: Matrix Scientific
047121-1G
047121-500MG
101878-530EA
1163.08
USD
101878-530
101878-528
2-[4-(tert-Butoxycarbonyl)piperazino]-3-chloroisonicotinic acid ≥95%
2-[4-(tert-Butoxycarbonyl)piperazino]-3-chloroisonicotinic acid
Hazard: Irritant
Formula:
C₁₅H₂₀ClN₃O₄ Melting Pt: 150…152 °C |
MDL Number:
MFCD11840992 CAS Number: 1135283-34-9 |
Specification Test Results
Melting point | 150-152°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...