2-(5-Chloro-2-pyridinyl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
047117-500MG
101878-526EA
375.38
USD
101878-526
2-(5-Chloro-2-pyridinyl)benzenecarboxylic acid ≥95%
2-(5-Chloro-2-pyridinyl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₈ClNO₂ Melting Pt: 193…195 °C |
MDL Number:
MFCD11100219 CAS Number: 1048912-89-5 |
Specification Test Results
Melting point | 193-195°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...