2-(5-Nitro-2-pyridinyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

047116-500MG
101878-524EA 375.38 USD
101878-524
2-(5-Nitro-2-pyridinyl)benzenecarboxylic acid ≥95%
2-(5-Nitro-2-pyridinyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₈N₂O₄
Melting Pt: 212…214 °C
MDL Number: MFCD11100218
CAS Number: 1048912-81-7

Order Now

Specification Test Results

Melting point 212-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR