(6-Chloro-3,4-dihydro-2H-1,4-benzoxazin-2-yl)methanol ≥95%
Supplier: Matrix Scientific
047114-500MG
047114-1G
101878-518EA
467.69
USD
101878-518
101878-516
(6-Chloro-3,4-dihydro-2H-1,4-benzoxazin-2-yl)methanol ≥95%
(6-Chloro-3,4-dihydro-2H-1,4-benzoxazin-2-yl)methanol
Hazard: Irritant
Formula:
C₉H₁₀ClNO₂ Melting Pt: 67…69 °C |
MDL Number:
MFCD11100217 CAS Number: 176383-57-6 |
Specification Test Results
Melting point | 67-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...