2-(4-Chlorobenzoyl)-3-(dimethylamino)acrylonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(4-Chlorophenyl)-2-[(dimethylamino)methylene]-3-oxopropanenitrile
047105-5G
047105-1G
047105-500MG
101878-484EA
500
USD
101878-484
101878-482
101878-480
2-(4-Chlorobenzoyl)-3-(dimethylamino)acrylonitrile ≥95%
2-(4-Chlorobenzoyl)-3-(dimethylamino)acrylonitrile
Hazard: Irritant
Formula:
C₁₂H₁₁ClN₂O MW: 234.69 g/mol Melting Pt: 110…112 °C |
MDL Number:
MFCD03453096 CAS Number: 52200-16-5 |
Specification Test Results
Melting point | 110-112°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...