3-(Dimethylamino)-2-(4-methylbenzoyl)acrylonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
2-[(Dimethylamino)methylene]-3-(4-methylphenyl)-3-oxopropanenitrile
047103-500MG
047103-1G
101878-476EA
698.46
USD
101878-476
101878-474
3-(Dimethylamino)-2-(4-methylbenzoyl)acrylonitrile ≥95%
3-(Dimethylamino)-2-(4-methylbenzoyl)acrylonitrile
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O MW: 214.27 g/mol Melting Pt: 132…134 °C |
MDL Number:
MFCD03453100 CAS Number: 96232-41-6 |
Specification Test Results
Melting point | 132-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...