6-Bromo-2-hydrazino-1,3-benzothiazole ≥95%
Supplier: Matrix Scientific
047093-5G
047093-1G
047093-500MG
101878-422EA
556.92
USD
101878-422
101878-426
101878-424
6-Bromo-2-hydrazino-1,3-benzothiazole ≥95%
6-Bromo-2-hydrazino-1,3-benzothiazole
Hazard: Irritant
Formula:
C₇H₆BrN₃S Melting Pt: 222…223 °C |
MDL Number:
MFCD04448804 CAS Number: 37390-63-9 |
Specification Test Results
Melting point | 222-223°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...