Methyl-2-ethyl-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

047058-1G 047058-500MG 047058-5G
101878-266EA 915.38 USD
101878-266 101878-264 101878-262
Methyl-2-ethyl-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-2-ethyl-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 44…45 °C
MDL Number: MFCD08689724
CAS Number: 924862-20-4

Order Now

Specification Test Results

Melting point 44-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR