2-Amino-6,8-dibromo-3-quinolinecarbonitrile ≥95%
Supplier: Matrix Scientific
047040-1G
047040-500MG
047040-5G
101878-166EA
806.15
USD
101878-166
101878-164
101878-162
2-Amino-6,8-dibromo-3-quinolinecarbonitrile ≥95%
2-Amino-6,8-dibromo-3-quinolinecarbonitrile
Hazard: Irritant
Formula:
C₁₀H₅Br₂N₃ MW: 326.99 g/mol Melting Pt: 313…315 °C |
MDL Number:
MFCD08689714 CAS Number: 937601-73-5 |
Specification Test Results
Melting point | 313-315°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...