3-(4-Bromophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047022-1G
047022-5G
047022-500MG
101878-076EA
532.31
USD
101878-076
101878-074
101878-072
3-(4-Bromophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-(4-Bromophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₄H₉BrN₂OS MW: 333.21 g/mol Melting Pt: 338…340 °C |
MDL Number:
MFCD02046230 CAS Number: 1028-39-3 |
Specification Test Results
Melting point | 338-340°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...