3-(3-Chloro-2-methylphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047018-500MG
047018-5G
047018-1G
101878-054EA
532.31
USD
101878-054
101878-052
101878-050
3-(3-Chloro-2-methylphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-(3-Chloro-2-methylphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₅H₁₁ClN₂OS MW: 302.78 g/mol Melting Pt: 286…288 °C |
MDL Number:
MFCD03460995 CAS Number: 81066-84-4 |
Specification Test Results
Melting point | 286-288°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...