5-Methyl-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047009-500MG
047009-5G
047009-1G
101878-014EA
532.31
USD
101878-014
101878-012
101878-010
5-Methyl-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
5-Methyl-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₅H₁₂N₂OS MW: 268.34 g/mol Melting Pt: 306…307 °C |
MDL Number:
MFCD08689695 CAS Number: 117106-06-6 |
Specification Test Results
Melting point | 306-307°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...