6,7-Dimethoxy-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-6,7-dimethoxy-3-phenyl-2-thioxo-1H-quinazolin-4-one

047008-5G 047008-1G 047008-500MG
101878-004EA 181.54 USD
101878-004 101878-008 101878-006
6,7-Dimethoxy-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
6,7-Dimethoxy-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₃S
MW: 314.37 g/mol
Melting Pt: 317…319 °C
MDL Number: MFCD03145974
CAS Number: 31485-66-2

Order Now

Specification Test Results

Melting point 317-319°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR