6,8-Dichloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
Synonyms:
6,8-Dichloro-2,3-dihydro-3-phenyl-2-thioxo-1H-quinazolin-4-one
047006-1G
047006-5G
047006-500MG
101877-998EA
181.54
USD
101877-998
101878-002
101878-000
6,8-Dichloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
6,8-Dichloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₄H₈Cl₂N₂OS MW: 323.2 g/mol Melting Pt: 274…275 °C |
MDL Number:
MFCD08689694 CAS Number: 67867-62-3 |
Specification Test Results
Melting point | 274-275°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...