5-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
Supplier: Matrix Scientific
047005-500MG
047005-5G
047005-1G
101877-992EA
181.54
USD
101877-992
101877-994
101877-996
5-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
5-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Hazard: Irritant
Formula:
C₁₄H₉ClN₂OS MW: 288.76 g/mol Melting Pt: 292…294 °C |
MDL Number:
MFCD08689693 CAS Number: 924869-18-1 |
Specification Test Results
Melting point | 292-294°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...