1-(tert-Butyl)-4-ethyl-5-(trifluoromethyl)-1H-pyrazole-1,4-dicarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Ethoxycarbonyl)-5-(trifluoromethyl)-1H-pyrazole, N-BOC protected
046999-500MG
046999-5G
046999-1G
101877-968EA
556.92
USD
101877-968
101877-970
101877-972
1-(tert-Butyl)-4-ethyl-5-(trifluoromethyl)-1H-pyrazole-1,4-dicarboxylate ≥95%
1-(tert-Butyl)-4-ethyl-5-(trifluoromethyl)-1H-pyrazole-1,4-dicarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₅F₃N₂O₄ MW: 308.26 g/mol Melting Pt: 107…108 °C |
MDL Number:
MFCD08689692 CAS Number: 1006492-02-9 |
Specification Test Results
Melting point | 107-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...