1-Benzyl-5-(hydroxymethyl)-6-(4-methoxyphenyl)tetrahydro-2(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
046998-500MG
046998-1G
046998-5G
101877-964EA
467.69
USD
101877-964
101877-966
101877-962
1-Benzyl-5-(hydroxymethyl)-6-(4-methoxyphenyl)tetrahydro-2(1H)-pyridinone ≥95%
1-Benzyl-5-(hydroxymethyl)-6-(4-methoxyphenyl)tetrahydro-2(1H)-pyridinone
Hazard: Irritant
Formula:
C₂₀H₂₃NO₃ Melting Pt: 80…82 °C |
MDL Number:
MFCD09027093 CAS Number: 937604-02-9 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...