2-Oxo-1,2-dihydro-4-pyrimidinecarbaldehyde oxime ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Oxo-1,2-dihydropyrimidine-4-carboxaldehyde oxime
046997-1G
046997-5G
046997-500MG
101877-956EA
278.46
USD
101877-956
101877-958
101877-960
2-Oxo-1,2-dihydro-4-pyrimidinecarbaldehyde oxime ≥95%
2-Oxo-1,2-dihydro-4-pyrimidinecarbaldehyde oxime
Hazard: Irritant
Formula:
C₅H₅N₃O₂ MW: 139.11 g/mol Melting Pt: 216 °C |
MDL Number:
MFCD02685605 CAS Number: 7460-56-2 |
Specification Test Results
Melting point | 216°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...