6-Bromo-2-chloro-3-quinolinecarbonitrile ≥95%
Supplier: Matrix Scientific
046988-1G
046988-500MG
101877-916EA
556.92
USD
101877-916
101877-918
6-Bromo-2-chloro-3-quinolinecarbonitrile ≥95%
6-Bromo-2-chloro-3-quinolinecarbonitrile
Hazard: Irritant
Formula:
C₁₀H₄BrClN₂ MW: 267.52 g/mol Melting Pt: 225…227 °C |
MDL Number:
MFCD09787668 CAS Number: 99465-04-0 |
Specification Test Results
Melting point | 225-227°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...